LIF372157894 Life Chemicals Inc.

1-(aminomethyl)cyclopentan-1-ol

  • MDL number MFCD09035274
  • Cas Number : 45511-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart