LIF372157895 Life Chemicals Inc.

[1-(pyrimidin-2-yl)piperidin-4-yl]methanamine

  • MDL number MFCD07365339
  • Cas Number : 158958-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart