LIF372157910 Life Chemicals Inc.

1-oxa-4-thia-8-azaspiro[4.5]decane

  • MDL number MFCD13176695
  • Cas Number : 5608-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart