LIF372157911 Life Chemicals Inc.

2-amino-1-(pyridin-2-yl)ethan-1-ol

  • MDL number MFCD03840171
  • Cas Number : 89943-14-6
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart