LIF372157915 Life Chemicals Inc.

(2-methoxyphenoxy)acetyl chloride

  • MDL number MFCD02223230
  • Cas Number : 40926-73-6
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart