LIF372157919 Life Chemicals Inc.

(2-methylphenoxy)acetyl chloride

  • MDL number MFCD11107992
  • Cas Number : 15516-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart