LIF372157920 Life Chemicals Inc.

(3-methylphenoxy)acetyl chloride

  • MDL number MFCD00235217
  • Cas Number : 40926-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart