LIF372157936 Life Chemicals Inc.

4-phenyltetrahydro-2H-pyran-4-carbonyl chloride

  • MDL number MFCD09903692
  • Cas Number : 100119-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart