LIF372157938 Life Chemicals Inc.

1-(chloroacetyl)piperidine

  • MDL number MFCD01095825
  • Cas Number : 1440-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart