LIF372157964 Life Chemicals Inc.

methyl 1-(4-aminophenyl)piperidine-4-carboxylate

  • MDL number MFCD16653413
  • Cas Number : 471938-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart