LIF372157966 Life Chemicals Inc.

4-(4-methoxypiperidin-1-yl)aniline

  • MDL number MFCD10034078
  • Cas Number : 1018635-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart