LIF372157999 Life Chemicals Inc.

N-(3-aminophenyl)cyclopropanesulfonamide

  • MDL number MFCD16653433
  • Cas Number : 1203416-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart