LIF372158020 Life Chemicals Inc.

1,3-diphenylprop-2-yn-1-one

  • MDL number MFCD00156767
  • Cas Number : 7338-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart