LIF372158021 Life Chemicals Inc.

2,2,4-Trimethyl-4-phenyl-1,2,3,4-tetrahydro-quinoline

  • MDL number MFCD00182056
  • Cas Number : 13562-24-8
  • Molecular Weight : 251.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart