LIF372158028 Life Chemicals Inc.

tert-butyl pent-4-en-1-ylcarbamate

  • MDL number MFCD08437624
  • Cas Number : 202925-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart