LIF372158035 Life Chemicals Inc.

2-{[(tert-butoxy)carbonyl]amino}-2-(oxan-4-yl)acetic acid

  • MDL number MFCD04114473
  • Cas Number : 182287-49-6
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart