LIF372158036 Life Chemicals Inc.

tert-butyl (3-hydroxycyclohexyl)carbamate

  • MDL number MFCD06658389
  • Cas Number : 610302-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart