LIF372158038 Life Chemicals Inc.

tert-butyl 3-(aminomethyl)morpholine-4-carboxylate

  • MDL number MFCD06738665
  • Cas Number : 475106-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart