LIF372158039 Life Chemicals Inc.

methyl 3-bromonaphthalene-1-carboxylate

  • MDL number MFCD06658478
  • Cas Number : 16650-63-8
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart