LIF372158040 Life Chemicals Inc.

6-fluoro-2-benzofuran-1(3H)-one

  • MDL number MFCD11053086
  • Cas Number : 23932-84-5
  • Molecular Weight : 152.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart