LIF372158042 Life Chemicals Inc.

2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD00039676
  • Cas Number : 573714-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart