LIF372158051 Life Chemicals Inc.

6-hydrazinylquinoline

  • MDL number MFCD09997909
  • Cas Number : 16023-69-1
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart