LIF372158053 Life Chemicals Inc.

6-methoxyquinolin-5-amine

  • MDL number MFCD12067881
  • Cas Number : 50358-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart