LIF372158058 Life Chemicals Inc.

1-[2-(1H-pyrazol-1-yl)ethyl]piperazine

  • MDL number MFCD14678763
  • Cas Number : 381721-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart