LIF372158060 Life Chemicals Inc.

[1-(2-hydroxyethyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD09046661
  • Cas Number : 926213-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart