LIF372158062 Life Chemicals Inc.

ethyl 5-bromopyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD06739182
  • Cas Number : 885276-93-7
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart