LIF372158065 Life Chemicals Inc.

methyl 2-aminoimidazo[1,2-a]pyridine-6-carboxylate

  • MDL number MFCD08059260
  • Cas Number : 885272-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart