LIF372158072 Life Chemicals Inc.

2-(5-amino-3-tert-butyl-1H-pyrazol-1-yl)-3,5,6,7-tetrahydro-4H-cyclopenta[d]pyrimidin-4-one

  • MDL number MFCD22587943
  • Cas Number : 1350989-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart