LIF372158073 Life Chemicals Inc.

1-(4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)-5-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD22587944
  • Cas Number : 1350988-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart