LIF372158082 Life Chemicals Inc.

2-(1-Oxo-1,2-dihydro-isoquinolin-5-yloxy)-propionic acid

  • MDL number MFCD14051628
  • Cas Number : 62000-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart