LIF372158083 Life Chemicals Inc.

5-methoxy-3,4-dihydroisoquinolin-1(2H)-one

  • MDL number MFCD07196047
  • Cas Number : 129075-49-6
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart