LIF372158089 Life Chemicals Inc.

2-[1-(2-hydroxyethyl)-4-oxo-1,4-dihydro-5H-pyrazolo[3,4-d]pyrimidin-5-yl]acetamide

  • MDL number MFCD07652846
  • Cas Number : 899971-27-8
  • Molecular Weight : 237.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart