LIF372158116 Life Chemicals Inc.

1-isobutyl-5-(4-methoxyphenyl)-1,3-dihydro-2H-imidazole-2-thione

  • MDL number MFCD16653449
  • Cas Number : 1105190-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart