LIF372158161 Life Chemicals Inc.

3-[(3-phenylpropanoyl)amino]benzoic acid

  • MDL number MFCD00423281
  • Cas Number : 446829-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart