LIF372158166 Life Chemicals Inc.

4-(3-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663861
  • Cas Number : 83558-37-6
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart