LIF372158168 Life Chemicals Inc.

2-(1H-benzimidazol-2-yl)ethanethiol

  • MDL number MFCD06009723
  • Cas Number : 2242-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart