LIF372158210 Life Chemicals Inc.

1-(3-hydroxyphenyl)-2,6-dimethylpyridin-4(1H)-one

  • MDL number MFCD06496382
  • Cas Number : 823176-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart