LIF372158223 Life Chemicals Inc.

1-(2-Fluorophenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD07397775
  • Cas Number : 887035-85-0
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-fluorophenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid
Inchi Code 1S/C10H8FN3O2/c1-6-9(10(15)16)12-13-14(6)8-5-3-2-4-7(8)11/h2-5H,1H3,(H,15,16)
Inchi Key HOCYRMQMIPAWLX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart