LIF372158234 Life Chemicals Inc.

6-chloro-N,N-dimethylpyridazin-3-amine

  • MDL number MFCD02732844
  • Cas Number : 7145-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart