LIF372158236 Life Chemicals Inc.

2-Methyl-5-oxo-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD03011582
  • Cas Number : 446829-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart