LIF372158267 Life Chemicals Inc.

N-benzyl-6-chloropyridazin-3-amine

  • MDL number MFCD00968535
  • Cas Number : 1496-85-1
  • Molecular Weight : 219.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-benzyl-6-chloro-3-pyridazinamine
Inchi Code 1S/C11H10ClN3/c12-10-6-7-11(15-14-10)13-8-9-4-2-1-3-5-9/h1-7H,8H2,(H,13,15)
Inchi Key HNJMQBUIQSWTSV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart