LIF372158289 Life Chemicals Inc.

methyl 3-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)propanoate

  • MDL number MFCD08143647
  • Cas Number : 90007-75-3
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl)propanoate
Inchi Code 1S/C8H10N2O4/c1-14-7(12)3-5-10-4-2-6(11)9-8(10)13/h2,4H,3,5H2,1H3,(H,9,11,13)
Inchi Key WFCIIRXTEXKYTF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart