LIF372158291 Life Chemicals Inc.

5-Methyl-4,5,6,7-tetrahydro-thiazolo[5,4-c]pyridin-2-ylamine

  • MDL number MFCD02854749
  • Cas Number : 17899-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart