LIF372158324 Life Chemicals Inc.

4-methoxy-2,5-dimethylbenzenesulfonyl chloride

  • MDL number MFCD08688025
  • Cas Number : 91179-12-3
  • Molecular Weight : 234.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart