LIF372158326 Life Chemicals Inc.

1-(cyclopropylcarbonyl)-1,2,3,4-tetrahydroquinolin-6-amine

  • MDL number MFCD09057596
  • Cas Number : 927966-09-4
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(cyclopropylcarbonyl)-1,2,3,4-tetrahydro-6-quinolinamine
Inchi Code 1S/C13H16N2O/c14-11-5-6-12-10(8-11)2-1-7-15(12)13(16)9-3-4-9/h5-6,8-9H,1-4,7,14H2
Inchi Key IFPFNXVWUVKXJT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart