LIF372158328 Life Chemicals Inc.

2-(2-oxopropyl)-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD00088346
  • Cas Number : 3416-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart