LIF372158345 Life Chemicals Inc.

ethyl 5-amino-3-methylthiophene-2-carboxylate

  • MDL number MFCD02257685
  • Cas Number : 88796-28-5
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-amino-3-methyl-2-thiophenecarboxylate
Inchi Code 1S/C8H11NO2S/c1-3-11-8(10)7-5(2)4-6(9)12-7/h4H,3,9H2,1-2H3
Inchi Key QMRFJZLWZKXXEH-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart