LIF372158348 Life Chemicals Inc.

6-Methoxypyrimidine-4-carboxylic acid

  • MDL number MFCD00496784
  • Cas Number : 38214-45-8
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart