LIF372158351 Life Chemicals Inc.

5-(p-tolyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD02672695
  • Cas Number : 33621-60-2
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-methylphenyl)-1,3,4-oxadiazol-2-amine
Inchi Code 1S/C9H9N3O/c1-6-2-4-7(5-3-6)8-11-12-9(10)13-8/h2-5H,1H3,(H2,10,12)
Inchi Key AXAUHVYAAUVJQR-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart