LIF372158364 Life Chemicals Inc.

{4-[(2,4-Dichlorobenzyl)oxy]phenyl}amine

  • MDL number MFCD08753566
  • Cas Number : 53397-81-2
  • Molecular Weight : 268.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart